Merck
Premature Drug Release from Polyethylene Glycol (PEG)-Coated Liposomal Doxorubicin via Formation of the Membrane Attack Complex.
ACS nano (2020-03-07)
Even Chen, Bing-Mae Chen, Yu-Cheng Su, Yuan-Chih Chang, Tian-Lu Cheng, Yechezekel Barenholz, Steve R Roffler
ABSTRAKT
Anti-polyethylene glycol (PEG) antibodies are present in many healthy individuals as well as in patients receiving polyethylene glycol-functionalized drugs. Antibodies against PEG-coated nanocarriers can accelerate their clearance, but their impact on nanodrug properties including nanocarrier integrity is unclear. Here, we show that anti-PEG IgG and IgM antibodies bind to PEG molecules on the surface of PEG-coated liposomal doxorubicin (Doxil, Doxisome, LC-101, and Lipo-Dox), resulting in complement activation, formation of the membrane attack complex (C5b-9) in the liposomal membrane, and rapid release of encapsulated doxorubicin from the liposomes. Drug release depended on both classical and alternative pathways of complement activation. Doxorubicin release of up to 40% was also observed in rats treated with anti-PEG IgG and PEG-coated liposomal doxorubicin. Our results demonstrate that anti-PEG antibodies can disrupt the membrane integrity of PEG-coated liposomal doxorubicin through activation of complement, which may alter therapeutic efficacy and safety in patients with high levels of pre-existing antibodies against PEG.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Dicyanocobinamide, ≥93%