Merck
Drought-induced lacuna formation in the stem causes hydraulic conductance to decline before xylem embolism in Selaginella.
The New phytologist (2020-05-10)
Amanda A Cardoso, Dominik Visel, Cade N Kane, Timothy A Batz, Clara García Sánchez, Lucian Kaack, Laurent J Lamarque, Yael Wagner, Andrew King, José M Torres-Ruiz, Déborah Corso, Régis Burlett, Eric Badel, Hervé Cochard, Sylvain Delzon, Steven Jansen, Scott A M McAdam
ABSTRAKT
Lycophytes are the earliest diverging extant lineage of vascular plants, sister to all other vascular plants. Given that most species are adapted to ever-wet environments, it has been hypothesized that lycophytes, and by extension the common ancestor of all vascular plants, have few adaptations to drought. We investigated the responses to drought of key fitness-related traits such as stomatal regulation, shoot hydraulic conductance (Kshoot ) and stem xylem embolism resistance in Selaginella haematodes and S. pulcherrima, both native to tropical understory. During drought stomata in both species were found to close before declines in Kshoot , with a 50% loss of Kshoot occurring at -1.7 and -2.5 MPa in S. haematodes and S. pulcherrima, respectively. Direct observational methods revealed that the xylem of both species was resistant to embolism formation, with 50% of embolized xylem area occurring at -3.0 and -4.6 MPa in S. haematodes and S. pulcherrima, respectively. X-ray microcomputed tomography images of stems revealed that the decline in Kshoot occurred with the formation of an air-filled lacuna, disconnecting the central vascular cylinder from the cortex. We propose that embolism-resistant xylem and large capacitance, provided by collapsing inner cortical cells, is essential for Selaginella survival during water deficit.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Auramine, analytical standard