Merck
A Decade of Varicella Screening Within a Paediatric Inflammatory Bowel Disease Population.
Journal of Crohn's & colitis (2020-01-01)
Rachel E Harris, Lee Curtis, Vikas Hegde, Vikki Garrick, Lisa Gervais, Lawrence Armstrong, Caroline Delahunty, Andrew Eccleston, Ghassan Al-Hourani, Diana M Flynn, Victoria Merrick, Andrew R Barclay, Rachel Tayler, Richard Hansen, Richard K Russell
ABSTRAKT
Increased risk of opportunistic infection-e.g., varicella zoster infection-secondary to therapies is a cause of morbidity in inflammatory bowel disease [IBD] patients. The UK vaccination schedule does not include varicella immunisation. We aimed to evaluate the varicella screening and immunisation programme in a paediatric IBD population. Data regarding IBD diagnosis, varicella status, and consequent immunisations/treatment interventions were collected retrospectively from the records of patients diagnosed with IBD over a 10-year period [2009-2018]. In all, 520 IBD patients were diagnosed; 505/520 [97%] had varicella testing; 46/505 [9%] were naïve. Of 501 patients, 391[78%] were tested before or within 7 days of diagnosis; this increased in the second 5-year period compared with the first (229/268 [85%] versus 162/233 [70%]; p <0.00001). Median diagnosis age of naïve patients was lower [8.3 years versus 12.8 years; p <0.00001]. Where vaccination was feasible, 21/31 [68%] had two and 7/31 [23%] one immunisation. Prednisolone induction led to lower rates of vaccination (5/13 [39%] versus 23/33 [70%] for other induction therapies; p =0.02). Of 28 vaccinated patients, 5 [18%] had suspected breakthrough varicella; and 6/18 [33%] unimmunised patients required post-exposure prophylaxis or treatment for varicella. Immunisation was associated with a decrease in patients requiring post-exposure prophylaxis (0/28 [0%] versus 5/18 [28%]; p =0.0006) and varicella-related hospital admission (1/28 [4%] versus 4/18 [22%]; p =0.01). High rates of varicella screening and immunisation within a PIBD population are possible, resulting in a reduction in hospital admissions for varicella treatment. Varicella immunisation may be of increasing importance within the PIBD population with the emergence of novel therapeutic strategies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Naphthoflavone, ≥98%