Merck
De novo design of peptides targeted to the EF hands of calmodulin.
The Journal of biological chemistry (2000-01-25)
M Villain, P L Jackson, M K Manion, W J Dong, Z Su, G Fassina, T M Johnson, T T Sakai, N R Krishna, J E Blalock
ABSTRAKT
This report describes the use of the concept of inversion of hydropathy patterns to the de novo design of peptides targeted to a predetermined site on a protein. Eight- and 12-residue peptides were constructed with the EF hands or Ca(2+)-coordinating sites of calmodulin as their anticipated points of interaction. These peptides, but not unrelated peptides nor those with the same amino acid composition but a scrambled sequence, interacted with the two carboxyl-terminal Ca(2+)-binding sites of calmodulin as well as the EF hands of troponin C. The interactions resulted in a conformational change whereby the 8-mer peptide-calmodulin complex could activate phosphodiesterase in the absence of Ca(2+). In contrast, the 12-mer peptide-calmodulin complex did not activate phosphodiesterase but rather inhibited activation by Ca(2+). This inhibition could be overcome by high levels of Ca(2+). Thus, it would appear that the aforementioned concept can be used to make peptide agonists and antagonists that are targeted to predetermined sites on proteins such as calmodulin.