Merck
  • Cobalt nanoparticles trigger ferroptosis-like cell death (oxytosis) in neuronal cells: Potential implications for neurodegenerative disease.
Cobalt nanoparticles trigger ferroptosis-like cell death (oxytosis) in neuronal cells: Potential implications for neurodegenerative disease.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-02-16)
Govind Gupta, Anda Gliga, Jonas Hedberg, Angela Serra, Dario Greco, Inger Odnevall Wallinder, Bengt Fadeel
ABSTRAKT
The neurotoxicity of hard metal-based nanoparticles (NPs) remains poorly understood. Here, we deployed the human neuroblastoma cell line SH-SY5Y differentiated or not into dopaminergic- and cholinergic-like neurons to study the impact of tungsten carbide (WC) NPs, WC NPs sintered with cobalt (Co), or Co NPs versus soluble CoCl2 . Co NPs and Co salt triggered a dose-dependent cytotoxicity with an increase in cytosolic calcium, lipid peroxidation, and depletion of glutathione (GSH). Co NPs and Co salt also suppressed glutathione peroxidase 4 (GPX4) mRNA and protein expression. Co-exposed cells were rescued by N-acetylcysteine (NAC), a precursor of GSH, and partially by liproxstatin-1, an inhibitor of lipid peroxidation. Furthermore, in silico analyses predicted a significant correlation, based on similarities in gene expression profiles, between Co-containing NPs and Parkinson's disease, and changes in the expression of selected genes were validated by RT-PCR. Finally, experiments using primary human dopaminergic neurons demonstrated cytotoxicity and GSH depletion in response to Co NPs and CoCl2 with loss of axonal integrity. Overall, these data point to a marked neurotoxic potential of Co-based but not WC NPs and show that neuronal cell death may occur through a ferroptosis-like mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade