Merck
Irisin, an exercise myokine, potently suppresses tumor proliferation, invasion, and growth in glioma.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-05-30)
Chiun-Wei Huang, Yu-Hsuan Chang, Hsuan-Hung Lee, Jing-Yi Wu, Jia-Xing Huang, Yi-Hsiu Chung, Shih-Ting Hsu, Lu-Ping Chow, Kuo-Chen Wei, Feng-Ting Huang
ABSTRAKT
Glioblastoma multiforme is the most common and aggressive glial tumor with poor prognosis. Importantly, effective treatment options for glioblastoma are unmet needs. Obesity and low physical activity have been linked with a high risk of cancer, and exercise is related to delayed cancer development and progression. Epidemiological studies have revealed a correlation between exercise and the survival rate of patients with glioblastoma. Nevertheless, the mechanisms by which exercise exerts its anticancer effects in glioblastoma remain unclear. Here, we found that irisin, an exercise-induced myokine, induced G2 /M cell cycle arrest and increased p21 levels in glioblastoma cells, leading to the inhibition of cell proliferation. In addition, irisin inhibited glioblastoma cell invasion by upregulating TFPI-2 and even reversed the aggressive tumor phenotype promoted by co-cultivation with cancer-associated adipocytes. Furthermore, irisin retarded xenograft glioblastoma tumor growth, and radiolabeled irisin demonstrated specific tumor-targeting capability in vivo. Therefore, this study identified one potential molecular mechanism by which exercise prevents cancer progression via irisin. Intriguingly, irisin has the potential to be developed as a molecular imaging and therapeutic anticancer agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human TFPI2