Merck
  • The MsrAB reducing pathway of Streptococcus gordonii is needed for oxidative stress tolerance, biofilm formation, and oral colonization in mice.
The MsrAB reducing pathway of Streptococcus gordonii is needed for oxidative stress tolerance, biofilm formation, and oral colonization in mice.
PloS one (2020-02-23)
Naif Jalal, Song F Lee
ABSTRAKT
The ability of Streptococcus gordonii to cope with oxidative stress is important for survival and persistence in dental plaque. In this study, we used mutational, phenotypic, and biochemical approaches to characterize the role of a methionine sulfoxide reductase (MsrAB) and proteins encoded by genes in the msrAB operon and an adjacent operon in oxidative stress tolerance in S. gordonii. The results showed that MsrAB and four other proteins encoded in the operons are needed for protection from H2O2 and methionine sulfoxide. These five proteins formed a reducing pathway that was needed for oxidative stress tolerance, biofilm formation, and oral colonization in mice. In the pathway, MsrAB was the enzyme that repaired oxidatively damaged proteins, and the two thioredoxin-like lipoproteins (SdbB and Sgo_1177) and two CcdA proteins were proteins that maintained the catalytic cycle of MsrAB. Consistent with the role in oxidative stress tolerance, the production of MsrAB, SdbB, and Sgo_11777 was induced in aerobic growth and planktonic cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Methionine sulfoxide
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile