Merck
Plasma concentrations of eleven cannabinoids in cattle following oral administration of industrial hemp (Cannabis sativa).
Scientific reports (2020-07-31)
Michael D Kleinhenz, Geraldine Magnin, Zhoumeng Lin, Jason Griffin, Katie E Kleinhenz, Shawnee Montgomery, Andrew Curtis, Miriam Martin, Johann F Coetzee
ABSTRAKT
Cannabinoid production for medicinal purposes has renewed interest in utilizing byproducts of industrial hemp (IH) as a feed source for livestock. However, the presence of bioactive residues in animal tissues may pose a risk to consumers. The purpose of this study was to characterize the plasma pharmacokinetics (PK) of cannabinoids and their metabolites in cattle after a single oral exposure to IH. Eight castrated male Holstein calves received a single oral dose of 35 g of IH to achieve a target dose of 5.4 mg/kg cannabidiolic acid (CBDA). Blood samples were collected for 96 h after dosing. Plasma cannabinoid concentrations were profiled using liquid chromatography coupled with mass-spectroscopy (UPLC) and PK parameters were calculated using noncompartmental methods. The cannabinoids CBDA, tetrahydrocannabinolic acid-A (THCA-A), cannabidivarinic acid (CBDVA), and cannabichromenic acid (CBCA) were detected in all cattle after IH dosing. The geometric mean maximum concentration of CBDA of 72.7 ng/mL was observed at 14 h after administration. The geometric mean half-life of CBDA was 14.1 h. No changes in serum biochemistry analysis were observed following IH dosing compared to baseline values. These results show acidic cannabinoids, especially CBDA, are readily absorbed from the rumen and available for distribution throughout the body.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cannabidiolic acid, analytical standard