Merck
  • Novel Steroidal 5α,8α-Endoperoxide Derivatives with Semicarbazone/Thiosemicarbazone Side-chain as Apoptotic Inducers through an Intrinsic Apoptosis Pathway: Design, Synthesis and Biological Studies.
Novel Steroidal 5α,8α-Endoperoxide Derivatives with Semicarbazone/Thiosemicarbazone Side-chain as Apoptotic Inducers through an Intrinsic Apoptosis Pathway: Design, Synthesis and Biological Studies.
Molecules (Basel, Switzerland) (2020-03-12)
Liwei Ma, Haijun Wang, Jing Wang, Lei Liu, Song Zhang, Ming Bu
ABSTRAKT
A series of novel steroidal 5α,8α-endoperoxide derivatives bearing semicarbazone (7a-g) or thiosemicarbazone (7h-k) side chain were designed, synthesized and evaluated for their cytotoxicities in four human cancer cell lines (HepG2, HCT-116, MCF-7, and A549) using the MTT assay in vitro. The results showed that compound 7j exhibited significant cytotoxic activity against HepG2 cells (IC50 = 3.52 μM), being more potent than ergosterol peroxide. Further cellular mechanism studies in HepG2 cells indicated that compound 7j triggered the mitochondrial-mediated apoptosis by decreasing mitochondrial membrane potential (MMP), which was associated with up-regulation of Bax, down-regulation of Bcl-2, activation levels of the caspase cascade, and formation of reactive oxygen species (ROS). The above findings indicated that compound 7j may be used as a promising skeleton for antitumor agents with improved efficacy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%