Merck
Effective inhibition of cancer cells by recombinant adenovirus expressing EGFR-targeting artificial microRNA and reversed-caspase-3.
PloS one (2020-08-04)
Maoxiao Yan, Jia Chen, Hua Jiang, Yuqiong Xie, Chunchun Li, Lihong Chen, Beibei Yang, Jiang Cao
ABSTRAKT
The EGFR-targeting cancer therapies are commonly facing drug resistance, mostly due to mutations. Gene therapy with artificial microRNA targeting EGFR conserved sequence may avoid such problem. In this study, we constructed a recombinant adenovirus expressing EGFR-targeting artificial microRNA and active revCASP3 (Ad-EC), under the control of tumor-specific SLPI promoter, and evaluated its inhibitory effect on HEP-2 cancer cells both in vitro and in vivo. MTT assay showed that cell growth inhibition rate at 72h was 44.0% in Ad-EC group at MOI 50, while the rate was 7.7% in the control virus Ad-GFP group and 3.6% in Cetuximab (500 μg/ml) group respectively. Flow cytometry analysis revealed the late apoptotic cells rate was 36.1% in Ad-EC group, significantly higher than 6.5% of Ad-GFP group (p < 0.001). When Ad-EC (MOI 50) was combined with CDDP (0.25 μg/ml), late apoptotic cells rate increased to 61.2%, significantly higher than each monotherapy group (P < 0.001). The real-time xCELLigence system recorded an effective cell growth inhibition in Ad-EC and CDDP groups, and more enhanced effect in Ad-EC plus CDDP group. Western blot revealed that Ad-EC could inhibit the activation of AKT pathway and ERK1/2 pathway, while Cetuximab had the AKT pathway over-activated. In vivo experiments with HEP-2 xenograft in nude mice confirmed the tumor inhibition in Ad-EC, CDDP and Ad-EC plus CDDP groups compared with PBS group (P < 0.01). Collectively, these data support the effective inhibition of cancer cells by this novel gene therapy strategy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%