Merck
Biomarkers for prediction of mortality in left-sided infective endocarditis.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2020-03-15)
Rinaldo F Siciliano, Danielle M Gualandro, Marcio Sommer Bittencourt, Milena Paixão, Fabiana Marcondes-Braga, Alexandre de Matos Soeiro, Célia Strunz, Ana Paula Pacanaro, Christian Puelacher, Flavio Tarasoutchi, Salvatore Di Somma, Bruno Caramelli, Mucio Tavares de Oliveira Junior, Alfredo Jose Mansur, Christian Mueller, Antonio Carlos Pereira Barretto, Tânia Mara Varejão Strabelli
ABSTRAKT
Evidence regarding biomarkers for risk prediction in patients with infective endocarditis (IE) is limited. We aimed to investigate the value of a panel of biomarkers for the prediction of in-hospital mortality in patients with IE. Between 2016 and 2018, consecutive IE patients admitted to the emergency department were prospectively included. Blood concentrations of nine biomarkers were measured at admission (D0) and on the seventh day (D7) of antibiotic therapy: C-reactive protein (CRP), sensitive troponin I (s-cTnI), procalcitonin, B-type natriuretic peptide (BNP), neutrophil gelatinase-associated lipocalin (NGAL), interleukin 6 (IL6), tumor necrosis factor α (TNF-α), proadrenomedullin, alpha-1-acid glycoprotein, and galectin 3. The primary endpoint was in-hospital mortality. Among 97 patients, 56% underwent cardiac surgery, and in-hospital mortality was 27%. At admission, six biomarkers were independent predictors of in-hospital mortality: s-cTnI (OR 3.4; 95%CI 1.8-6.4; P<0.001), BNP (OR 2.7; 95%CI 1.4-5.1; P=0.002), IL-6 (OR 2.06; 95%CI 1.3-3.7; P=0.019), procalcitonin (OR 1.9; 95%CI 1.1-3.2; P=0.018), TNF-α (OR 1.8; 95%CI 1.1-2.9; P=0.019), and CRP (OR 1.8; 95%CI 1.0-3.3; P=0.037). At admission, S-cTnI provided the highest accuracy for predicting mortality (area under the ROC curve: s-cTnI 0.812, BNP 0.727, IL-6 0.734, procalcitonin 0.684, TNF-α 0.675, CRP 0.670). After 7 days of antibiotic therapy, BNP and inflammatory biomarkers improved their performance (s-cTnI 0.814, BNP 0.823, IL-6 0.695, procalcitonin 0.802, TNF-α 0.554, CRP 0.759). S-cTnI concentration measured at admission had the highest accuracy for mortality prediction in patients with IE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%