Merck
Brain Imaging Study on the Pathogenesis of Depression & Therapeutic Effect of Selective Serotonin Reuptake Inhibitors.
Psychiatry investigation (2020-07-08)
Qi Meng, Aixia Zhang, Xiaohua Cao, Ning Sun, Xinrong Li, YunQiao Zhang, Yanfang Wang
ABSTRAKT
Predefining the most effective treatment for patients with depressive disorders remains a problem. We will examine the differential brain regions of gray matter (GM) in major depressive disorder (MDD) patients and the relationship between changes in their volume and the efficacy of early antidepressant treatment using magnetic resonance imaging (MRI). 159 never-medicated patients with first-episode MDD and 53 normal control subjects (NCs) were enrolled. The brains were scanned by MRI and measured with the 17-item Hamilton Depression Rating Scale (HAMD-17) at baseline and after 2 weeks of treatment with selective serotonin reuptake inhibitor (SSRI)s, and the non-responder group and responder group were obtained. The patients were analyzed by voxel-based morphological (VBM) and SPSS software. Receiver operator characteristics (ROC) analysis was performed for the difference between the responder group and the non-responder group in the differential brain regions, and Pearson correlations were computed between volume size and HAMD score reduction rate. Smaller GM volume of the right superior temporal gyrus (STG), and the orbital parts of the right medial frontal gyrus and right inferior frontal gyrus were observed in MDD versus the NCs. The non-responder group demonstrated a significant volume reduction at the right STG compared with the responders, but no corresponding change in orbital part of right medial frontal gyrus and right inferior frontal gyrus. ROC analysis showed that Accuracy=71.2%. There was a positive correlation between the STG gray matter volume and the HAMD-17 score reduction rate (r=0.347, p=0.002). The study results confirmed the local changes in brain structure in MDD and may initially predict the early treatment response produced by SSRIs as antidepressants.