Merck
  • Tumor suppressor OTUD3 induces growth inhibition and apoptosis by directly deubiquitinating and stabilizing p53 in invasive breast carcinoma cells.
Tumor suppressor OTUD3 induces growth inhibition and apoptosis by directly deubiquitinating and stabilizing p53 in invasive breast carcinoma cells.
BMC cancer (2020-06-24)
Qian Pu, Yan-Rong Lv, Ke Dong, Wen-Wen Geng, Hai-Dong Gao
ABSTRAKT
P53 pathway inactivation plays an important role in the process of breast cancer tumorigenesis. Post-translational protein modification abnormalities have been confirmed to be an important mechanism underlying inactivation of p53. Numerous deubiquitinating enzymes are aberrantly expressed in breast cancer, and a few deubiquitination enzymes can deubiquitinate and stabilize p53. Here, we report that ovarian tumor (OTU) deubiquitinase 3 (OTUD3) is a deubiquitylase of p53 in breast carcinoma (BC). Correlations between the mRNA expression levels of OTUD3, TP53 and PTEN and the prognosis of BC were assessed with the Kaplan-Meier Plotter tool. OTUD3 protein expression in 80 pairs of specimens in our cohort was examined by immunohistochemistry and western blotting. The relationship among OTUD3, p53, and p21 proteins was analyzed. Half-life analysis and ubiquitylation assay were performed to elucidate the molecular mechanism by which OTUD3 stabilizes p53. The interaction between OTUD3 and p53 in BC cells was verified by a co-immunoprecipitation assay and GST pulldown experiments. MTS assay for proliferation detection, detection of apoptosis induced by cisplatin and colony formation assay were employed to investigate the functional effects of OTUD3 on breast cancer cells. OTUD3 downregulation is correlated with a poor prognosis in BC patients. OTUD3 expression is decreased in breast cancer tissues and not associated with the histological grade. OTUD3 also inhibits cell proliferation and clone formation and increases the sensitivity of BC cells to apoptosis induced by chemotherapy drugs. Reduced OTUD3 expression accompanied by decreased p53 abundance is correlated with human breast cancer progression. Ectopic expression of wild-type OTUD3, but not its catalytically inactive mutant, stabilizes and activates p53. Mechanistically, OTUD3 interacts directly with p53 through the amino-terminal OTU region. Finally, OTUD3 protects p53 from murine double minute 2 (Mdm2)-mediated ubiquitination and degradation, enabling the deubiquitination of p53 in BC cells. In summary, we found that OTUD3 may be a potential therapeutic target for restoring p53 function in breast cancer cells and suggest that the OTUD3-p53 signaling axis may play a critical role in tumor suppression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)