Merck
  • The p48 Flow Modulation Device with Hydrophilic Polymer Coating (HPC) for the Treatment of Acutely Ruptured Aneurysms: Early Clinical Experience Using Single Antiplatelet Therapy.
The p48 Flow Modulation Device with Hydrophilic Polymer Coating (HPC) for the Treatment of Acutely Ruptured Aneurysms: Early Clinical Experience Using Single Antiplatelet Therapy.
Cardiovascular and interventional radiology (2020-02-08)
Marta Aguilar-Perez, Victoria Hellstern, Muhammad AlMatter, Christina Wendl, Hansjörg Bäzner, Oliver Ganslandt, Hans Henkes
ABSTRAKT
Flow diversion (FD) remains a potential treatment option following aneurysmal subarachnoid hemorrhage (aSAH) when standard options may not be feasible. However, it should not be considered a first-line treatment due to the need for dual antiplatelet therapy (DAPT). The hydrophilic polymer coating on the p48MW flow diverter (HPC, phenox) is a surface modification that inhibits platelet adhesion. This study aims to report on our early single-center experience using the p48MW HPC (phenox) flow diverter with single antiplatelet therapy (SAPT) following an aSAH. We retrospectively identified all patients who had been treated with the p48MW HPC for aSAH under SAPT. All patients treated within 30 days following an aSAH were included. Any occurrence of thromboembolic and hemorrhagic complications was recorded alongside angiographic and clinical follow-up details. Eight patients were identified. The mean interval between aSAH and FD was 6 days. Of the eight ruptured aneurysms, one was blister-like, one saccular, one mycotic, and the remaining five were dissecting aneurysms. Intraprocedural transient thrombus formation was observed in four patients (50%). Stent thrombosis was observed in one patient (12.5%) on day 3 with spontaneous recanalization after being switched onto DAPT. None of the aneurysms rebled after treatment. Two patients died due to cerebral vasospasm. Complete aneurysm occlusion had been achieved in all but one patient at angiographic follow-up (average 6 months). This small series highlights the possibility and limitations of using the p48MW HPC with SAPT in ruptured aneurysms. Randomized trials with longer follow-up in larger cohorts are underway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Prasugrel, ≥98% (HPLC)