Merck
Egg yolk-free cryopreservation of bull semen.
PloS one (2019-10-16)
Muhammad Anzar, Kosala Rajapaksha, Lyle Boswall
ABSTRAKT
Egg yolk is a common ingredient of mammalian semen extender to protect sperm against initial cold shock. However, egg yolk has biosecurity risks. Our main objectives were to cryopreserve bull semen without egg yolk using exogenous cholesterol and to study the protective role of glycerol in egg yolk-free semen extender. Other objectives were to compare protein profiles and in vitro fertilization potential of bull sperm frozen with and without egg yolk. In first experiment, semen was either diluted in conventional tris-egg yolk glycerol (TEYG control) extender or first treated with cholesterol-cyclodextrin complex (CC, 2 mg/ml semen) followed by dilution in egg yolk-free tris-glycerol (TG) extender (collectively called as "CC+TG") at 22°C or 4°C, and frozen. Post-thaw sperm motion characteristics were similar between CC+TG and TEYG control extenders, and temperature of glycerol addition. In second experiment, semen was frozen in CC+TG extender varying in glycerol concentration (7 to 0%; v/v). Post-thaw sperm quality decreased with the decline in glycerol concentration in TG extender, even higher concentration of CC complex (3 or 4 mg/ml semen) could not protect sperm in the absence of glycerol in TG extender. In third experiment, SDS electrophoresis of proteins from fresh sperm and sperm frozen in CC+TG, and TEYG control extenders was conducted. Protein profiles in fresh sperm and CC+TG frozen sperm were almost similar. Egg yolk proteins bound tightly with sperm plasma membrane. In fourth experiment, in vitro fertilization potentials of sperm frozen in TEYG control and CC+TG extenders were tested. Cleavage and blastocyst rates of semen frozen in CC+TG and TEYG control extenders were similar. In conclusion, cholesterol-cyclodextrin replaced egg yolk from the semen extender; glycerol remained essential for egg yolk-free sperm cryopreservation; and CC+TG extender did not modify sperm plasma membrane CC+TG whereas egg yolk extender changed the plasma membrane composition of bull sperm.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use