Merck
The metabolic changes in the hippocampus of an atherosclerotic rat model and the regulation of aerobic training.
Metabolic brain disease (2020-04-03)
Beibei Liu, Jingjing Li, Xiaojing Lin, Jingyun Hu, Shujie Lou
ABSTRAKT
Atherosclerosis has been associated with the progression of cognitive impairment and the effect of metabolic changes in the brain on cognitive function may be pronounced. The aim is to reveal the metabolic changes during atherosclerosis and clarify the possible role of exercise in regulating hippocampal metabolism. Hence, A rat model of atherosclerosis was established by high-fat diet feeding in combination with vitamin D3 intraperitoneal injection, then 4 weeks of aerobic exercise was conducted. Metabolomics based on GC-MS was applied to detect small molecules metabolites and western blot was used to detect the concentration of enzymes involved in metabolic changes in rat hippocampus. Compared to the control group, metabolites including xylulose 5-phosphate, threonine, succinate, and nonanoic acid were markedly elevated, whereas methyl arachidonic acid and methyl stearate decreased in the AS group, accompanied by a raised concentration of aldose reductase and glucose 6-phosphate dehydrogenase as well as a declined concentration of acetyl-CoA carboxylase and fatty acid synthase. After 4 weeks' aerobic exercise, the levels of succinic acid, branched chain amino acids, nonanoic acid, desmosterol, and aldose reductase decreased, whereas methyl arachidonic acid, methyl stearate, and glyceraldehyde-3-phosphate elevated in the hippocampus of the TAS group in comparison with the AS group. These results suggest that atherosclerosis could cause a severe metabolic disturbance, and aerobic exercise plays an important role in regulating atherosclerosis-induced disorder of glucose metabolism in the hippocampus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)