Merck
Active Notch signaling is required for arm regeneration in a brittle star.
PloS one (2020-05-13)
Vladimir Mashanov, Jennifer Akiona, Maleana Khoury, Jacob Ferrier, Robert Reid, Denis Jacob Machado, Olga Zueva, Daniel Janies
ABSTRAKT
Cell signaling pathways play key roles in coordinating cellular events in development. The Notch signaling pathway is highly conserved across all multicellular animals and is known to coordinate a multitude of diverse cellular events, including proliferation, differentiation, fate specification, and cell death. Specific functions of the pathway are, however, highly context-dependent and are not well characterized in post-traumatic regeneration. Here, we use a small-molecule inhibitor of the pathway (DAPT) to demonstrate that Notch signaling is required for proper arm regeneration in the brittle star Ophioderma brevispina, a highly regenerative member of the phylum Echinodermata. We also employ a transcriptome-wide gene expression analysis (RNA-seq) to characterize the downstream genes controlled by the Notch pathway in the brittle star regeneration. We demonstrate that arm regeneration involves an extensive cross-talk between the Notch pathway and other cell signaling pathways. In the regrowing arm, Notch regulates the composition of the extracellular matrix, cell migration, proliferation, and apoptosis, as well as components of the innate immune response. We also show for the first time that Notch signaling regulates the activity of several transposable elements. Our data also suggests that one of the possible mechanisms through which Notch sustains its activity in the regenerating tissues is via suppression of Neuralized1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
TRI Reagent®