Merck
PKM2 ablation enhanced retinal function and survival in a preclinical model of retinitis pigmentosa.
Mammalian genome : official journal of the International Mammalian Genome Society (2020-04-29)
Ethan Zhang, Joseph Ryu, Sarah R Levi, Jin Kyun Oh, Chun Wei Hsu, Xuan Cui, Ting-Ting Lee, Nan-Kai Wang, Jose Ronaldo Lima de Carvalho, Stephen H Tsang
ABSTRAKT
Retinitis pigmentosa (RP) is a neurodegenerative disorder that causes irreversible vision loss in over 1.5 million individuals world-wide. The genetic heterogeneity of RP necessitates a broad therapy that is able to provide treatment in a gene- and mutation- non-specific manner. In this study, we identify the therapeutic benefits of metabolic reprogramming by targeting pyruvate kinase M2 (PKM2) in a Pde6β preclinical model of RP. The genetic contributions of PKM2 inhibition in retinal degeneration were evaluated through histology and electroretinogram (ERG) followed by a statistical analysis using a linear regression model. Notably, PKM2 ablation resulted in thicker retinal layers in Pde6β-mutated mice as compared to the controls, suggesting greater photoreceptor survival. Consistent with these anatomical findings, ERG analyses revealed that the maximum b-wave is on average greater in Pkm2 knockout mice than in mice with intact Pkm2, indicating enhanced photoreceptor function. These rescue phenotypes from Pkm2 ablation in a preclinical model of RP indicate that a metabolome reprogramming may be useful in treating RP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution