Merck
Short-term angiotensin II treatment regulates cardiac nanomechanics via microtubule modifications.
Nanoscale (2020-07-29)
Pamela Swiatlowska, Jose L Sanchez-Alonso, Catherine Mansfield, Denis Scaini, Yuri Korchev, Pavel Novak, Julia Gorelik
ABSTRAKT
Mechanical properties of single myocytes contribute to the whole heart performance, but the measurement of mechanics in living cells at high resolution with minimal force interaction remains challenging. Angiotensin II (AngII) is a peptide hormone that regulates a number of physiological functions, including heart performance. It has also been shown to contribute to cell mechanics by inducing cell stiffening. Using non-contact high-resolution Scanning Ion Conductance Microscopy (SICM), we determine simultaneously cell topography and membrane transverse Young's modulus (YM) by a constant pressure application through a nanopipette. While applying pressure, the vertical position is recorded and a deformation map is generated from which YM can be calculated and corrected for the uneven geometry. High resolution of this method also allows studying specific membrane subdomains, such as Z-grooves and crests. We found that short-term AngII treatment reduces the transversal YM in isolated adult rat cardiomyocytes acting via an AT1 receptor. Blocking either a TGF-β1 receptor or Rho kinase abolishes this effect. Analysis of the cytoskeleton showed that AngII depletes microtubules by decreasing long-lived detyrosinated and acetylated microtubule populations. Interestingly, in the failing cardiomyocytes, which are stiffer than controls, the short-term AngII treatment also reduces the YM, thus normalizing the mechanical state of cells. This suggests that the short-term softening effect of AngII on cardiac cells is opposite to the well-characterized long-term hypertrophic effect. In conclusion, we generate a precise nanoscale indication map of location-specific transverse cortical YM within the cell and this can substantially advance our understanding of cellular mechanics in a physiological environment, for example in isolated cardiac myocytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder