Merck
MCM-22, MCM-36, and ITQ-2 Zeolites with Different Si/Al Molar Ratios as Effective Catalysts of Methanol and Ethanol Dehydration.
Materials (Basel, Switzerland) (2020-05-28)
Monika Marosz, Bogdan Samojeden, Andrzej Kowalczyk, Małgorzata Rutkowska, Monika Motak, Urbano Díaz, Antonio E Palomares, Lucjan Chmielarz
ABSTRAKT
MCM-22, MCM-36, and ITQ-2 zeolites with the intended Si/Al molar ratios of 15, 25, and 50 were synthetized and tested as catalysts for dehydration of methanol to dimethyl ether and dehydration of ethanol to diethyl ether and ethylene. The surface concentration of acid sites was regulated by the synthesis of zeolite precursors with different aluminum content in the zeolite framework, while the influence of porous structure on the overall efficiency of alcohol conversion was analyzed by application of zeolitic materials with different types of porosity-microporous MCM-22 as well as microporous-mesoporous MCM-36 and ITQ-2. The zeolitic samples were characterized with respect to their: chemical composition (ICP-OES), structure (XRD, FT-IR), texture (N2 sorption), and surface acidity (NH3-TPD). Comparison of the catalytic activity of the studied zeolitic catalysts with other reported catalytic systems, including zeolites with the similar Si/Al ratio as well as γ-Al2O3 (one of the commercial catalysts for methanol dehydration), shows a great potential of MCM-22, MCM-36, and ITQ-2 in the reactions of alcohols dehydration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N
Sigma-Aldrich
Hexamethyleneimine, 99%