Merck
Contamination of some foodstuffs marketed in Italy by fatty acid esters of monochloropropanediols and glycidol.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2020-02-23)
Eleonora Di Campi, Mauro Di Pasquale, Ettore Coni
ABSTRAKT
Fatty acid esters of 3-monochloropropanediol (3-MCPD), 2-MCPD and glycidol (Gly) are food-processing contaminants that cause concerns about possible adverse health effects. The present study evaluates the contamination levels of the three ester classes in 130 samples of foodstuffs marketed in Italy covering 10 food categories, namely margarines, oils, roasted coffee, breakfast cereals, salted crackers, cookies, infant biscuits, rusks, breads and potato crisps. The analytical method employed is a so-called indirect method that entails MCPD/glycidol cleavage from their esterified forms, cleanup, derivatisation and GC-MS analysis. The MCPDs and glycidol concentrations (from esters) were found to be equal or a little higher than the levels reported in previous studies conducted in other European countries and described in the literature. 3-MCPD was the predominant compound in all foodstuffs analysed with the exception for rusks where Gly levels were slightly higher. Considering the sum of MCPD and Gly esters, the most contaminated foodstuffs were seed oils, followed by margarines and cookies, whereas roasted coffee, bread, rusks, cornflakes and infant biscuits were less contaminated with MCPDs and Gly concentrations often below LOQ or LOD values. Refined olive oil, potato chips and salted crackers showed contamination levels intermediate between the two above groups. The results of this study also confirm that the use of palm oil as an ingredient or frying medium is an important cause of increase of the levels of MCPD and Gly esters, especially in salted crackers, rusks and potato crisps. Finally, the Italian intake of 3-MCPD due to the various foods analysed has been calculated and related to TDI. The MoE for Gly was also estimated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)