Merck
Effect of fungal co-cultures on ligninolytic enzyme activities, H2O2 production, and orange G discoloration.
Preparative biochemistry & biotechnology (2020-02-06)
Juana Lira-Pérez, Refugio Rodríguez-Vázquez, Wilberth Chan-Cupul
ABSTRAKT
In this study, the effects of Aspergillus niger in coculture with the basidiomycetes, Trametes versicolor, T. maxima, and Ganoderma spp., were studied to assess H2O2 production and laccase (Lac), Lignin Peroxidase (LiP), and manganese peroxidase (MnP) activities. The results indicated that maximum discoloration was of 97%, in the T. maxima and A. niger coculture, where the concentration of H2O2 was 5 mg/L and 6.3 mg/L in cultures without and with dye, respectively. These concentrations of H2O2 were 1.6- and 1.8-fold higher than monocultures of T. maxima (3.37 mg/L) and A. niger (3.87 mg/L), respectively. In the same coculture, the LiP and MnP enzyme activities also increased 12-fold, (from 0.08 U/mg to 0.99 U/mg), and 67-fold, (from 0.11 U/mg to 7.4 U/mg), respectively. The Lac activity increased 1.7-fold (from 13.46 U/mg to 24 U/mg). Further, a Box-Behnken experimental design indicated a 1.8-fold increase of MnP activity (from 7.4 U/mg to 13.3 U/mg). In addition, dye discoloration regression model obtained from the Box-Behnken experimental design showed a positively correlation with H2O2, (R2 = 0.58) and a negatively correlation with Lac activity (R2 = -0.7).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Orange G, certified by the BSC, Dye content 80 %