- A novel FTL mutation responsible for neuroferritinopathy with asymmetric clinical features and brain anomalies.
A novel FTL mutation responsible for neuroferritinopathy with asymmetric clinical features and brain anomalies.
Parkinsonism & related disorders (2014-06-08)
Sébastien Moutton, Patricia Fergelot, Jean-Marc Trocello, Violaine Plante-Bordeneuve, Nada Houcinat, Emilie Wenisch, Vincent Larue, Pierre Brugières, Fabienne Clot, Didier Lacombe, Benoit Arveiler, Cyril Goizet
PMID24907184
MATERIAŁY