Merck
Development (Cambridge, England) (2020-06-17)
Wael Noor El-Nachef, Marianne E Bronner
ABSTRAKT
The enteric nervous system (ENS) is essential for normal gastrointestinal function. Although the embryonic origin of enteric neurons from the neural crest is well established, conflicting evidence exists regarding postnatal enteric neurogenesis. Here, we address this by examining the origin of de novo neurogenesis in the post-embryonic zebrafish ENS. Although new neurons are added during growth and after injury, the larval intestine appears to lack resident neurogenic precursors or classical glia marked by sox10, plp1a, gfap or s100 Rather, lineage tracing with lipophilic dye or inducible Sox10-Cre suggests that post-embryonic enteric neurons arise from trunk neural crest-derived Schwann cell precursors that migrate from the spinal cord into the intestine. Furthermore, the 5-HT4 receptor agonist prucalopride increases enteric neurogenesis in normal development and after injury. Taken together, the results suggest that despite the lack of resident progenitors in the gut, post-embryonic enteric neurogenesis occurs via gut-extrinsic Schwann cell precursors during development and injury, and is promoted by serotonin receptor agonists. The absence of classical glia in the ENS further suggests that neural crest-derived enteric glia might have evolved after the teleost lineage.This article has an associated 'The people behind the papers' interview.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GR 113808, ≥98% (HPLC), solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Prucalopride, ≥98% (HPLC)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer