Merck
Dual-Responsive Nanotubes Assembled by Amphiphilic Dendrimers: Controlled Release and Crosslinking.
Materials (Basel, Switzerland) (2020-08-14)
Minghui Zhang, Hui Yang, Jiazhong Wu, Siyu Yang, Danfeng Yu, Xu Wu, Aiqing Ma, Keji Sun, Jinben Wang
ABSTRAKT
Although stimuli-responsive release systems have attracted great attention in medical applications, there has been no attempt at "precise" deep profile control based on such systems, which is greatly need to improve oil recovery. With this in mind, we provided a facile and simple strategy to prepare stimuli-responsive composite capsules of amphiphilic dendrimers-poly(styrene sulfonic acid) sodium/halloysite nanotubes (HNTs) via layer-by-layer (LbL) self-assembly technique, controlling the release crosslinking agent methenamine under different pH or salinity conditions. The release time of methenamine encapsulated in multilayer shells is about 40 h, which can be prolonged with the introduction of salt or shortened via the addition of acid, which accordingly induces the gelation of polyacrylamide (PAM) solutions, taking from a few hours to a dozen days. This study provided a novel approach for controllable release of chemical agents and controllable crosslinking of deep profiles in many application fields.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Hydroxynaphthalimide triflate, electronic grade, ≥99%