Merck
  • Optimization for allitol production from D-glucose by using immobilized glucose isomerase and recombinant E. coli expressing D-psicose-3-epimerase, ribitol dehydrogenase and formate dehydrogenase.
Optimization for allitol production from D-glucose by using immobilized glucose isomerase and recombinant E. coli expressing D-psicose-3-epimerase, ribitol dehydrogenase and formate dehydrogenase.
Biotechnology letters (2020-05-27)
Xin Wen, Huibin Lin, Yilin Ren, Can Li, Chengjia Zhang, Xin Song, Jianqun Lin, Jianqiang Lin
ABSTRAKT
To develop a method combining enzymatic catalysis and resting-cell biotransformation to produce allitol from low cost substrate D-glucose. The recombinant E. coli expressing D-psicose-3-epimerase (DPE), ribitol dehydrogenase (RDH) and formate dehydrogenase (FDH) for allitol production from D-fructose was constructed. The optimizations of the cell catalytic conditions and the cell cultivation conditions were made. Then, 63.4 g allitol L-1 was obtained from 100 g D-fructose L-1 in 4 h catalyzed by the recombinant E. coli cells. In order to decrease the substrate cost, D-glucose was used as the substrate instead of D-fructose and immobilized glucose isomerase was used to convert D-glucose into D-fructose. In order to simplify allitol production process from D-glucose, one-pot reaction using the mixed catalysts was used and the reaction conditions were optimized. Finally, 12.7 g allitol L-1 was obtained from 50 g D-glucose L-1 catalyzed by the mixed catalysts of immobilized glucose isomerase and the recombinant E. coli cells. Allitol can be efficiently produced from low cost substrate D-glucose by using the method combining enzymatic catalysis and resting-cell biotransformation, which is the first report.