Merck
  • DDAO Controlled Synthesis of Organo-Modified Silica Nanoparticles with Encapsulated Fluorescent Boron Dipyrrins and Study of Their Uptake by Cancerous Cells.
DDAO Controlled Synthesis of Organo-Modified Silica Nanoparticles with Encapsulated Fluorescent Boron Dipyrrins and Study of Their Uptake by Cancerous Cells.
Molecules (Basel, Switzerland) (2020-08-23)
Aleksandr A Goncharenko, Ilya A Tarasyuk, Yuriy S Marfin, Kirill V Grzhegorzhevskii, Albert R Muslimov, Andrey B Bondarenko, Maxim D Lebedev, Ilya A Kuz'min, Artur S Vashurin, Kirill V Lepik, Alexander S Timin, Evgeniy V Rumyantsev
ABSTRAKT
The design of cargo carriers with high biocompatibility, unique morphological characteristics, and capability of strong bonding of fluorescent dye is highly important for the development of a platform for smart imaging and diagnostics. In this paper, BODIPY-doped silica nanoparticles were prepared through a "one-pot" soft-template method using a sol-gel process. Several sol-gel precursors have been used in sol-gel synthesis in the presence of soft-template to obtain the silica-based materials with the most appropriate morphological features for the immobilization of BODIPY molecules. Obtained silica particles have been shown to be non-cytotoxic and can be effectively internalized into the cervical cancer cell line (HeLa). The described method of synthesis allows us to obtain silica-based carriers with an immobilized fluorescent dye that provide the possibility for real-time imaging and detection of these carriers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
N,N-Dimethyldodecylamine N-oxide solution, ~30% in H2O