Merck
Effects of beta3-adrenergic receptor activation on rat urinary bladder hyperactivity induced by ovariectomy.
The Journal of pharmacology and experimental therapeutics (2009-06-12)
F Aura Kullmann, Brian J Limberg, Debra E Artim, Mansi Shah, Thomas R Downs, Dan Contract, John Wos, Jan S Rosenbaum, William C de Groat
ABSTRAKT
Voiding dysfunctions, including increased voiding frequency, urgency, or incontinence, are prevalent in the postmenopausal population. Beta(3)-adrenergic receptor (beta(3)AR) agonists, which relax bladder smooth muscle, are being developed to treat these conditions. We utilized the rat ovariectomy (OVX) model to investigate the effect of ovarian hormone depletion on bladder function and the potential for beta(3)AR agonists to treat bladder hyperactivity in this setting. OVX increased voiding frequency and decreased bladder capacity by approximately 25% in awake rats and induced irregular cystometrograms in urethane-anesthetized rats. Reverse transcription-polymerase chain reaction revealed three betaARs subtypes (beta(1,2,3)) in bladder tissue, and immunostaining indicated beta(3)AR localization in urothelium and detrusor. Receptor expression was not different in OVX and SHAM rats. The beta(3)AR agonist selectivity of BRL37344 [(+/-)-(R(*),R(*))-[4-[2-[[2-(3-chlorophenyl)-2-hydroxyethyl]amino]propyl]phenoxy]acetic acid sodium hydrate], TAK-677 [(3-((2R)-(((2R)-(3-chlorophenyl)-2-hydroxyethyl)amino)propyl)-1H-indol-7-yloxy)acetic acid], and FK175 [acetic acid, 2-[[(8S)-8-[[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]-6,7,8,9-tetrahydro-5H-benzocyclohepten-2-yl]oxy], ethyl ester, hydrochloride] was confirmed by examining the relative potency for elevation of cAMP in CHOK1 cells overexpressing the various rat betaARs. Intravenous injection of each of the beta(3)AR agonists (0.1-500 microg/kg) in anesthetized rats decreased voiding frequency, bladder pressure, and amplitude of bladder contractions. In bladder strips, beta(3)AR agonists (10(-12)-10(-4) M) decreased baseline tone and reduced spontaneous contractions. BRL37344 (5 mg/kg) and TAK-677 (5 mg/kg) injected intraperitoneally in awake rats decreased voiding frequency by 40 to 70%. These effects were not altered by OVX. The results indicate that OVX-induced bladder dysfunction, including decreased bladder capacity and increased voiding frequency, is not associated with changes in beta(3)AR expression or the bladder inhibitory effects of beta(3)AR agonists. This suggests that beta(3)AR agonists should prove effective for the treatment of overactive bladder symptoms in the postmenopausal population.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cell Freezing Medium-DMSO 1×, sterile-filtered, suitable for cell culture
Supelco
Urethane, analytical standard
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)