Merck
DNA Micro-Disks for the Management of DNA-Based Data Storage with Index and Write-Once-Read-Many (WORM) Memory Features.
Advanced materials (Deerfield Beach, Fla.) (2020-07-30)
Yeongjae Choi, Hyung Jong Bae, Amos C Lee, Hansol Choi, Daewon Lee, Taehoon Ryu, Jinwoo Hyun, Seojoo Kim, Hyeli Kim, Suk-Heung Song, Kibeom Kim, Wook Park, Sunghoon Kwon
ABSTRAKT
DNA-based data storage has attracted attention because of its higher physical density of the data and longer retention time than those of conventional digital data storage. However, previous DNA-based data storage lacked index features and the data quality of storage after a single access was not preserved, obstructing its industrial use. Here, DNA micro-disks, QR-coded micro-sized disks that harbor data-encoded DNA molecules for the efficient management of DNA-based data storage, are proposed. The two major features that previous DNA-based data-storage studies could not achieve are demonstrated. One feature is accessing data items efficiently by indexing the data-encoded DNA library. Another is achieving write-once-read-many (WORM) memory through the immobilization of DNA molecules on the disk and their enrichment through in situ DNA production. Through these features, the reliability of DNA-based data storage is increased by allowing selective and multiple accession of data-encoded DNA with lower data loss than previous DNA-based data storage methods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
3-(Trimethoxysilyl)propyl acrylate, 92%, contains 100 ppm BHT as inhibitor