Merck
Familial Mediterranean fever mutations lift the obligatory requirement for microtubules in Pyrin inflammasome activation.
Proceedings of the National Academy of Sciences of the United States of America (2016-12-03)
Hanne Van Gorp, Pedro H V Saavedra, Nathalia M de Vasconcelos, Nina Van Opdenbosch, Lieselotte Vande Walle, Magdalena Matusiak, Giusi Prencipe, Antonella Insalaco, Filip Van Hauwermeiren, Dieter Demon, Delfien J Bogaert, Melissa Dullaers, Elfride De Baere, Tino Hochepied, Joke Dehoorne, Karim Y Vermaelen, Filomeen Haerynck, Fabrizio De Benedetti, Mohamed Lamkanfi
ABSTRAKT
Familial Mediterranean fever (FMF) is the most common monogenic autoinflammatory disease worldwide. It is caused by mutations in the inflammasome adaptor Pyrin, but how FMF mutations alter signaling in FMF patients is unknown. Herein, we establish Clostridium difficile and its enterotoxin A (TcdA) as Pyrin-activating agents and show that wild-type and FMF Pyrin are differentially controlled by microtubules. Diverse microtubule assembly inhibitors prevented Pyrin-mediated caspase-1 activation and secretion of IL-1β and IL-18 from mouse macrophages and human peripheral blood mononuclear cells (PBMCs). Remarkably, Pyrin inflammasome activation persisted upon microtubule disassembly in PBMCs of FMF patients but not in cells of patients afflicted with other autoinflammatory diseases. We further demonstrate that microtubules control Pyrin activation downstream of Pyrin dephosphorylation and that FMF mutations enable microtubule-independent assembly of apoptosis-associated speck-like protein containing a caspase recruitment domain (ASC) micrometer-sized perinuclear structures (specks). The discovery that Pyrin mutations remove the obligatory requirement for microtubules in inflammasome activation provides a conceptual framework for understanding FMF and enables immunological screening of FMF mutations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution