Merck
CRISP3 expression drives prostate cancer invasion and progression.
Endocrine-related cancer (2020-05-02)
Marianna Volpert, Luc Furic, Jinghua Hu, Anne E O'Connor, Richard J Rebello, Shivakumar Keerthikumar, Jemma Evans, D Jo Merriner, John Pedersen, Gail P Risbridger, Peter McIntyre, Moira K O'Bryan
ABSTRAKT
Identifying the factors stimulating prostate cancer cells migration and invasion has the potential to bring new therapeutic targets to the clinic. Cysteine-rich secretory protein 3 (CRISP3) is one of the most highly upregulated proteins during the transition of a healthy human prostatic epithelium to prostate cancer. Here we show using a genetically engineered mouse model of prostate cancer that CRISP3 production greatly facilitates disease progression from carcinoma in situ to invasive prostate cancer in vivo. This interpretation was confirmed using both human and mouse prostate cancer cell lines, which showed that exposure to CRISP3 enhanced cell motility and invasion. Further, using mass spectrometry, we show that CRISP3 induces changes in abundance of a subset of cell-cell adhesion proteins, including LASP1 and TJP1 both in vivo and in vitro. Collectively, these data identify CRISP3 as being pro-tumorigenic in the prostate and validate it as a potential target for therapeutic intervention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human CRISP3
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, liquid, BioReagent, suitable for cell culture