Merck
Effects of fluorescent carbon dots from the baked lamb on energy and lipid metabolism.
Food chemistry (2020-08-21)
Lijuan Zhang, Xiaokang Na, Bin Lai, Yukun Song, Haitao Wang, Mingqian Tan
ABSTRAKT
Food-borne carbon dots (CDs) may cause health risks due to their unique properties. However, previous efforts were mainly focused on the characterization of their physicochemical properties, their effects on cellular metabolism are not entirely revealed. Herein, the features and potential toxicity of CDs from lamb baked for 15, 30, and 45 min were evaluated, their cytotoxicity increased with the extension of baking time. Furthermore, the metabolic responses of PC12 cells after exposure to CDs from lamb baked for 45 min were investigated. The CDs perturbed purine metabolism, causing reactive oxygen species accumulation. Meanwhile, the CDs down-regulated glycolysis and TCA cycle, led to a significant decrease in ATP. Additionally, the CDs induced triglyceride accumulation, mainly through enhanced fatty acid biosynthesis. The adverse effects of CDs from baked lamb involved the perturbation of energy production, purine metabolism, and triglyceride biosynthesis, which provided additional information about the risks of CDs from food items.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
JC-1, powder or solid (Crystals)