Merck
  • Competitive inhibitions of the chlorophyll synthase of Synechocystis sp. strain PCC 6803 by bacteriochlorophyllide a and the bacteriochlorophyll synthase of Rhodobacter sphaeroides by chlorophyllide a.
Competitive inhibitions of the chlorophyll synthase of Synechocystis sp. strain PCC 6803 by bacteriochlorophyllide a and the bacteriochlorophyll synthase of Rhodobacter sphaeroides by chlorophyllide a.
Journal of bacteriology (2009-11-03)
Eui-Jin Kim, Jeong K Lee
ABSTRAKT
The photosynthetic growth of Synechocystis sp. strain PCC 6803 is hampered by exogenously added bacteriochlorophyllide a (Bchlide a) in a dose-dependent manner. The growth inhibition caused by Bchlide a, however, is relieved by an increased level of exogenously added chlorophyllide a (Chlide a). The results are explained by the competitive inhibition of chlorophyll synthase by Bchlide a, with inhibition constants (K(I)s) of 0.3 mM and 1.14 mM in the presence of sufficient geranylgeranyl pyrophosphate (GGPP) and phytyl pyrophosphate (PPP), respectively. Surprisingly, the bacteriochlorophyll synthase of Rhodobacter sphaeroides is inhibited competitively by Chlide a, with K(I)s of 0.54 mM and 0.77 mM in the presence of sufficient GGPP and PPP, respectively. Consistently, exogenously added Chlide a inhibits the metabolic conversion of exogenously added Bchlide a to bacteriochlorophyll a by an R. sphaeroides bchFNB-bchZ mutant that neither synthesizes nor metabolizes Chlide a. The metabolic inhibition by Chlide a, however, is relieved by the elevated level of Bchlide a. Thus, the chlorophyll synthase of Synechocystis sp. PCC 6803 and the bacteriochlorophyll synthase of R. sphaeroides, both of which perform ping-pong-type reactions, are inhibited by Bchlide a and Chlide a, respectively. Although neither inhibitor is catalyzed by the target enzyme, inhibitions in the competitive mode suggest a structural similarity between their active sites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
CHL, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sigma-Aldrich
Phytol, 95%, mixture of isomers
Sigma-Aldrich
Phytol, ≥97%, FG