Merck
  • Associations of non-pedunculated T1 colorectal adenocarcinoma outcome with consensus molecular subtypes, immunoscore, and microsatellite status: a multicenter case-cohort study.
Associations of non-pedunculated T1 colorectal adenocarcinoma outcome with consensus molecular subtypes, immunoscore, and microsatellite status: a multicenter case-cohort study.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2020-06-26)
Krijn J C Haasnoot, Yara Backes, Leon M G Moons, Onno Kranenburg, Anne Trinh, Louis Vermeulen, Michaël Noë, Jurriaan B Tuynman, Anja U G van Lent, Rosaline van Ginneken, Cornelis A Seldenrijk, Mihaela G Raicu, Kari Trumpi, Inge Ubink, Anya N Milne, Jurjen J Boonstra, John N Groen, Matthijs P Schwartz, Frank H J Wolfhagen, Joost M J Geesing, Frank Ter Borg, Lodewijk A A Brosens, Jeroen van Bergeijk, Bernhard W M Spanier, Wouter H de Vos Tot Nederveen Cappel, Koen Kessels, Tom C J Seerden, Frank P Vleggaar, G Johan A Offerhaus, Peter D Siersema, Sjoerd G Elias, Miangela M Laclé
ABSTRAKT
Advanced colorectal cancer (CRC) consensus molecular subtype 4 (CMS4) or CRC with a low immunoscore is associated with shorter survival times. Non-metastatic CRC with microsatellite instability (MSI) is associated with a lower risk of recurrence. We evaluated outcome (lymph node metastases [LNM] or cancer recurrence) in these tumor subtypes in patients with surgically-removed non-pedunculated T1 CRC by performing a multicenter case-cohort study. We included all patients in 13 hospitals in the Netherlands from 2000-2014 (n = 651). We randomly selected a subgroup of patients (n = 223) and all patients with LNM or recurrence (n = 63), and median follow-up of 44 months. We centrally reviewed tumor-slides, and constructed and immunostained tissue microarrays determining MSI, CMS (MSI/CMS1, CMS2/3, or CMS4), and immunoscore (I-low/I-high). We used weighted Cox proportional hazard models to evaluate the association of MSI, CMS, and immunoscore with LNM or recurrence, adjusting for conventional histologic risk factors. In the randomly selected subgroup of patients, 7.1% of tumors were MSI/CMS1, 91.0% CMS2/3, 1.8% CMS4, and 25% I-low. In the case-cohort, patients with CMS4 tumors had an increased risk for LNM or recurrence compared with patients with tumors of other CMSs (adjusted hazard ratio [HR], 3.97; 95% CI, 1.12-14.06; P = 0.03). Albeit not significant, tumors with MSI had a lower risk for LNM or recurrence than other tumor subtypes (adjusted HR, 0.52; 95% CI, 0.12-2.30; P = 0.39), whereas tumors with a low immunoscore had an increased risk for LNM or recurrence (adjusted HR, 1.30; 95% CI, 0.68-2.48; P = 0.43). In conclusion, in a case-cohort study of patients with non-pedunculated T1 CRC, MSI, and immunoscore were not significantly associated with adverse outcome after surgery. CMS4 substantially increased the risk of adverse outcome. However, CMS4 is rare in T1 CRCs, limiting its value for determining the risk in patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-ZEB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HTR2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution