Merck
Rheological Investigation of Relaxation Behavior of Polycarbonate/Acrylonitrile-Butadiene-Styrene Blends.
Polymers (2020-08-29)
Jae Sik Seo, Ho Tak Jeon, Tae Hee Han
ABSTRAKT
The rheological properties of polycarbonate/acrylonitrile-butadiene-styrene (PC/ABS) blends with various blend ratios are investigated at different temperatures to determine the shear dependent chain motions in a heterogeneous blend system. At low frequency levels under 0.1 rad/s, the viscosity of the material with a blend ratio of 3:7 (PC:ABS) is higher than that of pure ABS polymer. As the temperature increases, the viscosities of ABS-rich blends increase rather than decrease, whereas PC-rich blends exhibit decrease in viscosity. Results from the time sweep measurements indicate that ordered structures of PC and the formation and breakdown of internal network structures of ABS polymer occur simultaneously in the blend systems. Newly designed sequence test results show that the internal structures formed between PC and ABS polymers are dominant at low shear conditions for the blend ratio of 3:7 and effects of structural change and the presence of polybutadiene (PBD) become dominant at high shear conditions for pure ABS. The results of yield stress and relaxation time for PC/ABS blends support this phenomenon. The specimen with a blend ratio of 3:7 exhibited the highest value of yield stress at high temperature among others, which implies that the internal structure become stronger at higher temperature. The heterogeneity of ABS-rich blends increases whereas that of PC-rich blends decreases as temperature increases.