Merck
An optimized quantitative proteomics method establishes the cell type-resolved mouse brain secretome.
The EMBO journal (2020-09-22)
Johanna Tüshaus, Stephan A Müller, Evans Sioma Kataka, Jan Zaucha, Laura Sebastian Monasor, Minhui Su, Gökhan Güner, Georg Jocher, Sabina Tahirovic, Dmitrij Frishman, Mikael Simons, Stefan F Lichtenthaler
ABSTRAKT
To understand how cells communicate in the nervous system, it is essential to define their secretome, which is challenging for primary cells because of large cell numbers being required. Here, we miniaturized secretome analysis by developing the "high-performance secretome protein enrichment with click sugars" (hiSPECS) method. To demonstrate its broad utility, hiSPECS was used to identify the secretory response of brain slices upon LPS-induced neuroinflammation and to establish the cell type-resolved mouse brain secretome resource using primary astrocytes, microglia, neurons, and oligodendrocytes. This resource allowed mapping the cellular origin of CSF proteins and revealed that an unexpectedly high number of secreted proteins in vitro and in vivo are proteolytically cleaved membrane protein ectodomains. Two examples are neuronally secreted ADAM22 and CD200, which we identified as substrates of the Alzheimer-linked protease BACE1. hiSPECS and the brain secretome resource can be widely exploited to systematically study protein secretion and brain function and to identify cell type-specific biomarkers for CNS diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), agarose conjugate, Type VI, saline suspension
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC50 = 15 nM for human BACE-1).
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Human CD200 ELISA Kit, for serum, plasma, cell culture supernatant and urine