Merck
  • SUMO-fusion and autoinduction-based combinatorial approach for enhanced production of bioactive human interleukin-24 in Escherichia coli.
SUMO-fusion and autoinduction-based combinatorial approach for enhanced production of bioactive human interleukin-24 in Escherichia coli.
Applied microbiology and biotechnology (2020-10-03)
Sana Tahir, M Mudassir Iqbal, M Waheed Akhtar, Qingbing Wang, Tao Sun, Saima Sadaf
ABSTRAKT
High-level production of recombinant human interleukin-24 (IL-24), a multifunctional immunomodulatory cytokine, has been challenging due primarily to its aggregation as inclusion bodies in the bacterial host while persistent poor-expression in the insect/mammalian expression systems. The present study presents a robust, vector-host combination (pE-SUMO-IL24), auto-inducible medium (YNG/M9NG), and a simple purification scheme for soluble, bioactive, and cost-effective production of native-like IL-24 (nIL-24) in Escherichia coli. The final protein yield, following a three-step purification scheme (IMAC, SEC, dialysis), was 98 mg/L in shake-flask culture (with scale-up potential), which was several folds higher than reported earlier. In vitro cytotoxicity assays with HeLa and HCT116 cancer cell lines (performed using different concentrations of nIL-24) and the fluorescence activated cell sorting analysis (FACS) revealed a dose- and concentration-dependent increase in the population of pro-apoptotic cells with concomitant, statistically significant drop in the number of cells existent at Go/G1-, S-, and G2/M-phases (P < 0.002). The bioactive nIL-24, developed through this study, holds promise for use in further functional characterizations/applications. KEY POINTS: • Yeast SUMO fusion partner at N-terminus for improved solubility of an otherwise insoluble IL-24 in E. coli. • Enhanced cell densities with concomitant several-fold increase in protein yield by lactose-inducible media. • Improved inhibition of cervical and colorectal carcinomas by native-like nIL-24 compared with Met-containing IL. • Heterologous nIL-24 may enable better understanding of the functional intricacies linked up with its unique cancer-specific features. Graphical abstract.