Merck
  • Simultaneous degradation of triazophos, methamidophos and carbofuran pesticides in wastewater using an Enterobacter bacterial bioreactor and analysis of toxicity and biosafety.
Simultaneous degradation of triazophos, methamidophos and carbofuran pesticides in wastewater using an Enterobacter bacterial bioreactor and analysis of toxicity and biosafety.
Chemosphere (2020-10-30)
Yuxiao Zhang, Zixiao Xu, Zhengjun Chen, Gejiao Wang
ABSTRAKT
Triazophos (TAP), methamidophos (MAP) and carbofuran (CF) pesticides are highly toxic, soluble and absorbable. Efficient co-degradation of multi-pesticides is rare reported. The objectives of this study were to investigate TAP, MAP and CF co-degradative ability of Enterobacter sp. Z1 and study the degradation mechanisms. Strain Z1 was shown to efficiently co-degrade TAP, MAP and CF when they were used as primary carbon sources. The degradation occurred over a wide range of temperatures, pH values and pesticide concentrations and followed first-order kinetics. Under the optimum conditions (37 °C, pH 7 and 100 mg/L of each pesticide), the degradation efficiencies were 100%, 100%, and 95.3% for TAP, MAP and CF, respectively. In addition, strain Z1 could simultaneously degrade TAP, MAP, CF and total nitrogen in wastewater in a batch bioreactor, with high removal efficiencies of 98.3%, 100%, 98.7% and 100%, respectively. Genomics, proteomics, qRT-PCR and gene overexpression analyses revealed that the degradation mechanisms involved the activities of multiple proteins, among which, organophosphorus hydrolase (Oph) and 3-hydroxyacyl-CoA dehydrogenase (PaaC) are primarily responsible for TAP and MAP degradation, while carbofuran hydrolase (Mcd) and amidohydrolase (RamA) primarily degrade CF. Among these enzymes, PaaC and RamA are newly identified pesticide-degrading enzymes. Toxicity assays of strain Z1 using reporter recombinase gene (recA) and zebrafish showed that there was no accumulation of toxic metabolites during the degradation process. Biosafety test using zebrafish showed that the strain was nontoxic toward zebrafish. Strain Z1 provides a good purification effect for pesticides-containing wastewater and novel microbial pesticide-degrading mechanisms were discovered.