Merck
Engineering of a Core-Shell Nanoplatform to Overcome Multidrug Resistance via ATP Deprivation.
Advanced healthcare materials (2020-09-19)
Genhua Liu, Liucan Wang, Junjie Liu, Lu Lu, Dong Mo, Ke Li, Xin Yang, Rui Zeng, Jixi Zhang, Peng Liu, Kaiyong Cai
ABSTRAKT
Inhibiting the function of P-glycoprotein (P-gp) transporter, which causes drug efflux through adenosine triphosphate (ATP)-dependent manner, has become an effective strategy to conquer multidrug resistance (MDR) of cancer cells. However, there remains challenges for effective co-delivery, sequential release of P-gp modulator and chemotherapeutic agent. In this work, a novel type of core-shell nanoparticle is reported. It can independently encapsulate a high amount (about 683 µg mg-1 ) of chemotherapeutic agent doxorubicin (DOX) in the mesoporous polydopamine (MPDA) core and glucose oxidase (GOx) in the zeolite imidazolate frameworks-8 (ZIF-8) shell, namely MPDA@ZIF-8/DOX+GOx. The fast release of GOx triggered by acid-sensitive degradation of the ZIF-8 shell consumes glucose to starve cancer cells for ATP deprivation and effective suppress ATP-dependent drug efflux in advance, and then effectively facilitates the accumulation of DOX in MCF-7/ADR cancer cells. Experiments in vitro and in vivo demonstrate that the fabricated nanosystem can dramatically improve anticancer effects for MDR through sequential release property and exhibit excellent biocompatibility. Overall, this work reveals new insights in the use of GOx for MDR treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.