Merck
  • MicroRNA-145 suppresses osteogenic differentiation of human jaw bone marrow mesenchymal stem cells partially via targeting semaphorin 3A.
MicroRNA-145 suppresses osteogenic differentiation of human jaw bone marrow mesenchymal stem cells partially via targeting semaphorin 3A.
Connective tissue research (2019-07-16)
Yucui Jin, Fangling Hong, Qianyi Bao, Qiufan Xu, Rui Duan, Zhu Zhu, Wei Zhang, Changyan Ma
ABSTRAKT
Purpose: Human jaw bone marrow mesenchymal stem cells (h-JBMMSCs) are multipotent progenitor cells with osteogenic differentiation potential. MicroRNAs (miRNAs) have emerged as crucial modulators of osteoblast differentiation. In this study, we focus on the role of miR-145 and its target protein in osteoblast differentiation of h-JBMMSCs. Materials and Methods: h-JBMMSCs were isolated and cultured in osteogenic medium. miR-145 mimics and inhibitors were used to elevate and inhibit miR-145 expression, respectively. Osteogenic differentiation was determined by Alkaline phosphatase (ALP) and Alizarin red S (ARS) staining, and osteogenic marker detection using quantitative real-time reverse transcription PCR (qRT-PCR) assay. Bioinformatic analysis and luciferase reporter assay were used to identify the target gene of miR-145. Results: MiR-145 was down-regulated during osteogenesis of h-JBMMSCs. Inhibition of miR-145 promoted osteogenic differentiation of h-JBMMSCs, revealed by enhanced activity of alkaline phosphatase (ALP), greater mineralisation, and increased expression levels of the osteogenic markers, such as Runt-related transcription factor 2 (RUNX2), Osterix (OSX), ALP and COL1A1. MiR-145 could negatively regulate semaphorin3A (SEMA3A), which acts as a positive regulator of osteogenesis. MiR-145 inhibitor induced osteogenesis could be partially attenuated by SEMA3A siRNA treatment in h-JBMMSCs. Conclusions: Our data show that miR-145 directly targets SEMA3A, and also suggest miR-145 as a suppressor, plays an important role in the osteogenic differentiation of h-JBMMSCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human SEMA3A