Merck
Bioprinting small diameter blood vessel constructs with an endothelial and smooth muscle cell bilayer in a single step.
Biofabrication (2020-07-04)
Lei Xu, Mathew Varkey, Adam Jorgensen, Jihui Ju, Qianheng Jin, Ji Hoon Park, Yi Fu, Guangliang Zhang, Dongxu Ke, Weixin Zhao, Ruixing Hou, Anthony Atala
ABSTRAKT
Bioengineered artificial blood vessels have been a major area of interest over the last decade. Of particular interest are small diameter vessels, as surgical options are currently limited. This study aimed to fabricate a small diameter, heterogeneous bilayer blood vessel-like construct in a single step with gelatin methacryloyl (GelMA) bioink using a 3D micro-extrusion bioprinter on a solid platform. GelMA was supplemented with Hyaluronic acid (HA), glycerol and gelatin to form a GelMA bioink with good printability, mechanical strength, and biocompatibility. Two separate concentrations of GelMA bioink with unique pore sizes were selected to fabricate a heterogeneous bilayer. A higher concentration of GelMA bioink (6% w/v GelMA, 2% gelatin, 0.3% w/v HA, 10% v/v glycerol) was used to load human umbilical vein endothelial cells (HUVECs) and form an inner, endothelial tissue layer. A lower concentration of GelMA bioink (4% w/v GelMA, 4% gelatin, 0.3% w/v HA, 10% v/v glycerol) was used to load smooth muscle cells (SMCs) and form an outer, muscular tissue layer. Bioprinted blood vessel-like grafts were then assessed for mechanical properties with Instron mechanical testing, and suture-ability, and for biological properties including viability, proliferation, and histological analysis. The resulting 20 mm long, 4.0 mm diameter lumen heterogeneous bilayer blood vessel-like construct closely mimics a native blood vessel and maintains high cell viability and proliferation. Our results represent a novel strategy for small diameter blood vessel biofabrication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
SAFC
DMEM/Nutrient Mixture F-12 Ham, With 15 mM HEPES, without L-glutamine, L-leucine, L-lysine, L-methionine, CaCl2, MgCl2, MgSO4, sodium bicarbonate, and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A