Merck
Investigation of Biofilms Formed on Steelmaking Slags in Marine Environments for Water Depuration.
International journal of molecular sciences (2020-09-26)
Akiko Ogawa, Reiji Tanaka, Nobumitsu Hirai, Tatsuki Ochiai, Ruu Ohashi, Karin Fujimoto, Yuka Akatsuka, Masanori Suzuki
ABSTRAKT
Steelmaking slags are a promising resource as artificial seaweed beds for the reconstitution of marine environments. To grow seaweed well, the formation of biofilms is an essential process in biofouling. This study focused on the formation of initial biofilms on steelmaking slag samples and analyzed the resulting bacterial communities using the next-generation sequencing technique. Three types of steelmaking slag were submerged in an area of Ise Bay in Mie Prefecture, Japan, for 3 and 7 days in the summer and winter seasons to allow the formation of biofilms. The bacterial communities of these biofilms were richer in sulfur-oxidizing bacteria compared to the biofilms formed on polyurethane sponges. It was found that Helicobacteraceae dominantly grew on the biofilms formed on the slag samples. This shows that steelmaking slags have potential to be used as artificial seaweed beds and marine water purifiers.