Merck
  • Carbonic anhydrase inhibitors. Synthesis and inhibition of cytosolic/tumor-associated carbonic anhydrase isozymes I, II, and IX with boron-containing sulfonamides, sulfamides, and sulfamates: toward agents for boron neutron capture therapy of hypoxic tumors.
Carbonic anhydrase inhibitors. Synthesis and inhibition of cytosolic/tumor-associated carbonic anhydrase isozymes I, II, and IX with boron-containing sulfonamides, sulfamides, and sulfamates: toward agents for boron neutron capture therapy of hypoxic tumors.
Bioorganic & medicinal chemistry letters (2005-05-24)
Jean-Yves Winum, Alessandro Cecchi, Jean-Louis Montero, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRAKT
A library of boron-containing carbonic anhydrase (CA, EC 4.2.1.1) inhibitors, including sulfonamides, sulfamides, and sulfamates is reported. The new compounds have been synthesized by derivatization reactions of 4-carboxy-/amino-/hydroxy-phenylboronic acid pinacol esters with amino/isothiocyanato-substituted aromatic/heteroaromatic sulfonamides or by sulfamoylation reactions with sulfamoyl chloride. The new derivatives have been assayed for the inhibition of three physiologically relevant CA isozymes, the cytosolic CA I and II, and the transmembrane, tumor-associated isozyme CA IX. Effective inhibitors were detected both among sulfonamides, sulfamates, and sulfamides. Against the human isozyme hCA I the new compounds showed inhibition constants in the range of 34-94nM, against hCA II in the range of 3.1-48nM, and against hCA IX in the range of 7.3-89nM, respectively. As hypoxic tumors highly overexpress CA IX, the design of boron-containing inhibitors with high affinity for the tumor-associated CA isozymes may lead to important advances in boron neutron capture therapy (BNCT) applications targeting such tumors, which are non-responsive to both classical chemo- and radiotherapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)