Merck
  • Pentacyclic triterpenes. Part 1: the first examples of naturally occurring pentacyclic triterpenes as a new class of inhibitors of glycogen phosphorylases.
Pentacyclic triterpenes. Part 1: the first examples of naturally occurring pentacyclic triterpenes as a new class of inhibitors of glycogen phosphorylases.
Bioorganic & medicinal chemistry letters (2005-09-20)
Xiaoan Wen, Hongbin Sun, Jun Liu, Guanzhong Wu, Luyong Zhang, Xiaoming Wu, Peizhou Ni
ABSTRAKT
The semi-synthesis, in vitro and in vivo biological evaluation of corosolic acid (1) and maslinic acid (2) are described. Compounds 1 and 2 represent a new class of inhibitors of glycogen phosphorylases. Both 1 and 2 inhibit the increase of fasted plasma glucose of diabetic mice induced by adrenaline. It is therefore proposed that naturally occurring pentacyclic triterpenes 1 and 2 might reduce blood glucose, at least in part, through inhibiting hepatic glycogen degradation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, BioXtra