Merck
Furocoumarin glycosides from the leaves of Ficus ruficaulis Merr. var. antaoensis.
Journal of natural products (2005-02-01)
Mau-Sun Chang, Yuh-Cheng Yang, Yuh-Chi Kuo, Yueh-Hsiung Kuo, Chen Chang, Chin-Mei Chen, Tzong-Huei Lee
ABSTRAKT
From the methanolic extract of the leaves of Ficus ruficaulis Merr. var. antaoensis, 5-O-beta-D-glucopyranosyl-6-hydroxyangelicin (1), 6-O-beta-D-glucopyranosyl-5-hydroxyangelicin (2), 5,6-O-beta-D-diglucopyranosylangelicin (3), 8-O-beta-D-glucopyranosyl-5-hydroxypsoralen (4), 5-O-beta-D-glucopyranosyl-8-hydroxypsoralen (5), 3,4,5-trihydroxydehydro-alpha-ionol-9-O-beta-D-glucopyranoside (6), rutin (7), and isoquercitrin (8) were isolated. The structures of compounds 1-4 were elucidated by the analysis of their spectroscopic data. Their in vitro antiproliferation activities were also evaluated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid