Merck
Lysine-Based Biodegradable Surfactants: Increasing the Lipophilicity of Insulin by Hydrophobic Ion Paring.
Journal of pharmaceutical sciences (2020-08-08)
Markus Kurpiers, Julian Dominik Wolf, Helen Spleis, Christian Steinbring, Arne Matteo Jörgensen, Barbara Matuszczak, Andreas Bernkop-Schnürch
ABSTRAKT
The aim of this study was to evaluate biodegradable cationic surfactants based on lysine. Lysine was esterified with cholesterol, oleyl alcohol and 1-decanol resulting in cholesteryl lysinate (CL), oleyl lysinate (OL) and decyl lysinate (DL). Esters were investigated regarding their log Dn-octanol/water, critical micelle concentration (CMC) and biodegradability. Hemolytic potential of CL, OL, DL and the already established hexadecyl lysinate (HL) was determined and complexes with insulin (INS) were formed by hydrophobic ion pairing (HIP). Lipophilic characteristics of ion-pairs were examined by analyzing their log Pn-butanol/water. Successful synthesis of CL, OL and DL was confirmed by IR, NMR and MS. Log D analysis revealed amphiphilic properties for the esters and a CMC of 0.01 mM, 2.0 mM and 6.0 mM was found for CL, OL and DL, respectively. Biodegradability was proven, as over 99% of OL and DL were degraded by isolated enzymes within 30 min and after 3 h 97% of CL was cleaved by membrane bound enzymes. OL as well as DL displayed no hemolytic effect and for CL cytotoxicity was significantly reduced in comparison to HL. INS/CL complex exhibited highest lipophilicity. Cholesterol-amino acid based surfactants seem to be promising agents for HIP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%
Sigma-Aldrich
Triton X-100, laboratory grade