Merck
Structural Basis for the Binding Selectivity of Human CDY Chromodomains.
Cell chemical biology (2020-05-30)
Cheng Dong, Yanli Liu, Tian-Jie Lyu, Serap Beldar, Kelsey N Lamb, Wolfram Tempel, Yanjun Li, Zoey Li, Lindsey I James, Su Qin, Yun Wang, Jinrong Min
ABSTRAKT
The CDY (chromodomain on the Y) proteins play an essential role in normal spermatogenesis and brain development. Dysregulation of their expression has been linked to male infertility and various neurological diseases. Like the chromodomains of HP1 and Polycomb, the CDY chromodomains also recognize the lysine-methylated ARKS motif embedded in histone and non-histone proteins. Interestingly, the CDY chromodomains exhibit different binding preferences for the lysine-methylated ARKS motif in different sequence contexts. Here, we present the structural basis for selective binding of CDY1 to H3K9me3 and preferential binding of CDYL2 to H3tK27me3 over H3K27me3. In addition, we use a CDYL1/2-selective compound, UNC4850, to gain further insight into the molecular mechanisms underlying CDYL2 binding specificity. Our work also provides critical implications that CDYL1b's role in the regulation of neural development is dependent on its recognition of the lysine-methylated ARKS motif.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
UNC4850, ≥95% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)