Merck
Rapid synthesis of triazole-modified resveratrol analogues via click chemistry.
Journal of medicinal chemistry (2006-01-20)
Francesca Pagliai, Tracey Pirali, Erika Del Grosso, Riccardo Di Brisco, Gian Cesare Tron, Giovanni Sorba, Armando A Genazzani
ABSTRAKT
Resveratrol is a phytoalexin able to display an array of biological activities. We decided to replace the double bond with a triazole ring using the archetypical click reaction: the Huisgen [3 + 2] cycloaddition. Seventy-two triazole derivatives were synthesized via a parallel combinatorial approach. Preliminary data suggest that this procedure can lead to the synthesis of compounds that display some, but not all, of resveratrol's actions with increased potency.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)