- Synthesis and biological investigation of new 4''-malonyl tethered derivatives of erythromycin and clarithromycin.
Synthesis and biological investigation of new 4''-malonyl tethered derivatives of erythromycin and clarithromycin.
Bioorganic & medicinal chemistry letters (2006-01-03)
Daniel Sherman, Liqun Xiong, Alexander S Mankin, Artem Melman
PMID16387493
ABSTRAKT
A new approach to 4''-substituted derivatives of erythromycin and clarithromycin was developed by converting them into corresponding 4''-malonic monoesters. Subsequent carbodiimide coupling with alcohols and amines provided new macrolide derivatives that are capable of binding to 50S ribosomal subunits and inhibiting protein synthesis in cell-free system.