Merck
Novel dual inhibitors of calpain and lipid peroxidation with enhanced cellular activity.
Bioorganic & medicinal chemistry letters (2005-12-29)
Serge Auvin, Bernadette Pignol, Edith Navet, Morgane Troadec, Denis Carré, José Camara, Dennis Bigg, Pierre-E Chabrier
ABSTRAKT
A series of dipeptides with dual inhibitory activities on calpain and lipid peroxidation were prepared to target the intracellular calpain. This optimization program focused on the variations of the linker and the N-terminal amino acid of the peptidic core. Two compounds 6d-05 and 6d-08 exhibited potent intracellular calpain inhibition. The polar surface area and the number of rotors appeared to be critical descriptors to account for the behavior of these hybrid molecules in the cellular calpain assay.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder